2-Methylbenzylamine, 96%
2-Methylbenzylamine, 96%
2-Methylbenzylamine, 96%
Thermo Scientific Chemicals

2-Methylbenzylamine, 96%

CAS: 89-93-0 | C8H11N | 121.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 155630050
Price (EUR)
90,30
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
90,30
Each
Chemical Identifiers
CAS89-93-0
IUPAC Name1-(2-methylphenyl)methanamine
Molecular FormulaC8H11N
InChI KeyCJAAPVQEZPAQNI-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1CN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Titration with HClO4>=95.0 %
Infrared spectrumConforms
Refractive index1.5415 to 1.5435 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only