Di-2-pyridyl ketone, 98%
Di-2-pyridyl ketone, 98%
Di-2-pyridyl ketone, 98%
Thermo Scientific Chemicals

Di-2-pyridyl ketone, 98%

CAS: 19437-26-4 | C11H8N2O | 184.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 155670050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS19437-26-4
IUPAC Name2-(pyridine-2-carbonyl)pyridine
Molecular FormulaC11H8N2O
InChI KeyQPOWUYJWCJRLEE-UHFFFAOYSA-N
SMILESO=C(C1=CC=CC=N1)C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to beige to brown
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals and/or chunks
GC>=97.5 %
Melting point49°C to 54°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only