Di-2-piridil cetona, 98 %, Thermo Scientific Chemicals
Di-2-piridil cetona, 98 %, Thermo Scientific Chemicals
Di-2-piridil cetona, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Di-2-piridil cetona, 98 %, Thermo Scientific Chemicals

CAS: 19437-26-4 | C11H8N2O | 184.2 g/mol
Have Questions?
Número de catálogoCantidadEnvase
15567025025 gFrasco de vidrio
Número de catálogo 155670250
Precio (USD)
-
Cantidad:
25 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS19437-26-4
IUPAC Name2-(pyridine-2-carbonyl)pyridine
Molecular FormulaC11H8N2O
InChI KeyQPOWUYJWCJRLEE-UHFFFAOYSA-N
SMILESO=C(C1=CC=CC=N1)C1=CC=CC=N1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Light yellow to beige to brown
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals and/or chunks
GC>=97.5 %
Melting point49°C to 54°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only