UltraPure™ Ethidium Bromide, 10 mg/mL
Invitrogen™

UltraPure™ Ethidium Bromide, 10 mg/mL

UltraPure™ Ethidium Bromide is a sensitive fluorescent dye used to detect nucleic acids in agarose gels and Caesium chloride (CsCl)Read more
Have Questions?
Catalog NumberQuantity
1558501110 mL
Catalog number 15585011
Price (EUR)
93,75
Each
Quantity:
10 mL
Request bulk or custom format
Price (EUR)
93,75
Each
UltraPure™ Ethidium Bromide is a sensitive fluorescent dye used to detect nucleic acids in agarose gels and Caesium chloride (CsCl) gradients. It is available as an aqueous solution containing 10 mg/ml UltraPure™ Ethidium Bromide and high-purity water.
• Sensitive: Detect as little as little as 1 ng of nucleic acids in agarose gels
• Versatile: Detect DNA or RNA
• Easy to Use: Can be used as-is or diluted to the desired concentration

UltraPure™ Ethidium Bromide is an aromatic compound with a phenanthridine core (see structure below). Ethidium bromide has excitation maxima at 300 nm and 520 nm and an emission maximum at 600 nm (see fluorescence spectra).

For research use only. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection LocationIn-Gel Detection
Detection MethodFluorescence
Quantity10 mL
Shipping ConditionRoom Temperature
Target MoleculeRNA, DNA
Label or DyeEthidium Bromide
Product LineUltraPure
Product TypeEthidium Bromide
Unit SizeEach
Contents & Storage
10ml of 10mg⁄ml solution. Store at 15°C to 30°C

Frequently asked questions (FAQs)

Do you have a protocol for ethidium bromide staining in agarose gels?

For use in agarose gels:

- Add ethidium bromide to melted agarose to a final concentration of 0.5 µg/mL. Do not melt agarose that already contains ethidium bromide.

For staining agarose gels after electrophoresis:

- You can stain gels that have been run in the absence of ethidium bromide by covering the gel in 0.5 µg/mL ethidium bromide in water and gently agitating for 10 to 30 minutes.
- If necessary, gels can be destained by shaking in H2O for an additional 30 minutes. To stain RNA gels, you should minimize staining time and definitely include a destaining period.

What can ethidium bromide be used to detect and what is its sensitivity?

Ethidium bromide can be used to detect ssDNA, RNA, and dsDNA. This fluorescent dye intercalates between the stacked bases of nucleic acids, and exhibits an increased fluorescence at 590 nm. Ethidium bromide can detect down to ~1-10 ng/band of dsDNA in an agarose gel.

What amount of ethidium bromide do I need to stain nucleic acids in gels?

After electrophoresis, the gel should be stained in a 0.5-1.0 µg/mL solution of ethidium bromide in deionized water for 15 to 60 min depending on the thickness of the gel. As an optional step to reduce background fluorescence, the gel can be destained in deionized water for 15 to 30 min. Alternatively, ethidium bromide may be added directly to the agarose prior to casting. Add ethidium bromide to melted agarose to a final concentration of 0.5 µg/mL. This has the advantage of reducing the amount of ethidium bromide waste. However, this procedure may reduce the migration rate of nucleic acids.

Citations & References (2)

Citations & References
Abstract
Immune-like phagocyte activity in the social amoeba.
Authors:Chen G, Zhuchenko O, Kuspa A,
Journal:Science
PubMed ID:17673666
Social amoebae feed on bacteria in the soil but aggregate when starved to form a migrating slug. We describe a previously unknown cell type in the social amoeba, which appears to provide detoxification and immune-like functions and which we term sentinel (S) cells. S cells were observed to engulf bacteria ... More
Cloning and pharmacological characterization of the guinea pig P2X7 receptor orthologue.
Authors:Fonfria E, Clay WC, Levy DS, Goodwin JA, Roman S, Smith GD, Condreay JP, Michel AD,
Journal:Br J Pharmacol
PubMed ID:18037910
BACKGROUND AND PURPOSE: The human, rat, and mouse P2X(7) receptors have been previously characterized, and in this study we report the cloning and pharmacological properties of the guinea pig orthologue. EXPERIMENTAL APPROACH: A cDNA encoding for the guinea pig P2X(7) receptor was isolated from a guinea pig brain library. The ... More