Diethylamine hydrochloride, 99%, Thermo Scientific Chemicals
Diethylamine hydrochloride, 99%, Thermo Scientific Chemicals
Diethylamine hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethylamine hydrochloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 155891000
Price (EUR)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS660-68-4
IUPAC Namediethylazanium chloride
Molecular FormulaC4H12ClN
InChI KeyHDITUCONWLWUJR-UHFFFAOYSA-N
SMILES[Cl-].CC[NH2+]CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Melting point227°C to 230°C
Loss on drying=<1 %
Titration with HClO4>=98.5 %
Appearance (Color)White to off-white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only