Diethylamine hydrochloride, 99%, Thermo Scientific Chemicals
Diethylamine hydrochloride, 99%, Thermo Scientific Chemicals
Diethylamine hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethylamine hydrochloride, 99%, Thermo Scientific Chemicals

数量:
100 g
500 g
1 kg
5 kg
パッケージ:
すべて表示
Plastic bottle
製品番号(カタログ番号) 155895000
価格(JPY)
-
数量:
500 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS660-68-4
IUPAC Namediethylazanium chloride
Molecular FormulaC4H12ClN
InChI KeyHDITUCONWLWUJR-UHFFFAOYSA-N
SMILES[Cl-].CC[NH2+]CC
さらに表示
Appearance (Form)Crystalline powder or crystals
Melting point227°C to 230°C
Loss on drying=<1 %
Titration with HClO4>=98.5 %
Appearance (Color)White to off-white
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only