UltraPure™ SSPE, 20X
Invitrogen™

UltraPure™ SSPE, 20X

UltraPure™ 20X SSPE is a prepared buffer concentrate formulated for use in nucleic acid hybridizations and blot transfer applications. SuppliedRead more
Have Questions?
Catalog NumberQuantity
155910431 L
Catalog number 15591043
Price (USD)
106.00
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
106.00
Each

UltraPure™ 20X SSPE is a prepared buffer concentrate formulated for use in nucleic acid hybridizations and blot transfer applications. Supplied in 1 L plastic bottles, or in a 4 L or 10 L stackable Cubitainer¤ Box, sterile-filtered UltraPure™ 20X SSPE contains 3.0 M NaCl, 0.2 M NaH2PO4, and 0.02 M EDTA at pH 7.4.

Performance and Quality Testing: No DNase or RNase activity detected.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration20 X
For Use With (Application)Nucleic Acid Gel Electrophoresis, Blotting
FormLiquid
Product LineUltraPure
Product TypeSSPE
Quantity1 L
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store at room temperature.