UltraPure™ Phenol:Chloroform:Isoamyl Alcohol (25:24:1, v/v)
Invitrogen™

UltraPure™ Phenol:Chloroform:Isoamyl Alcohol (25:24:1, v/v)

UltraPure™ Phenol:Chloroform:Isoamyl Alcohol (25:24:1, v/v) is used in the purification of nucleic acids. This reagent consists of highly pure chloroform,Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
15593031100 mL
15593049400 mL
Catalog number 15593031
Price (USD)
194.65
Online exclusive
203.00
Save 8.35
Each
Quantity:
100 mL
Price (USD)
194.65
Online exclusive
203.00
Save 8.35
Each

UltraPure™ Phenol:Chloroform:Isoamyl Alcohol (25:24:1, v/v) is used in the purification of nucleic acids. This reagent consists of highly pure chloroform, isoamyl alcohol, and UltraPure™ Phenol saturated with Tris-HCl.

Using Phenol:Chloroform:Isoamyl Alcohol
When mixtures are extracted with Phenol:Chloroform:Isoamyl Alcohol, proteins are denatured and collected in the organic phase or at the interphase, while nucleic acids remain in the aqueous phase. UltraPure™ Phenol:Chloroform:lsoamyl Alcohol contains no preservatives. It is packaged under an inert gas in shatter-resistant, plastic-coated amber bottles.

Performance and quality testing
The appearance of the solution is evaluated at room temperature. No RNase or DNase activity is detected.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialPhenols
Product LineUltraPure
PurityMolecular Biology Grade
Quantity100 mL
Shipping ConditionRoom Temperature
FormLiquid
pH8.05
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C. To extend product stability, it is recommended that the gas volume above the solution be flushed with argon or nitrogen following each use.

Frequently asked questions (FAQs)

Do you have any information on DNA and RNA purification using phenol chloroform and alcohol precipitation?

Phenol extraction of proteins:

Phenol extraction is frequently used to remove proteins from nucleic acid solutions. A common protocol is to add an equal volume of buffer-saturated phenol or phenol:chloroform:isoamyl alcohol (25:24:1, v/v/v) to an aqueous nucleic acid solution, vortex, and centrifuge at 14,000 x g for 1 min to separate the phases.

Studies at Thermo Fisher Scientific have shown that the concentration of NaCl in the aqueous solution should not exceed 0.5 M for good recovery of DNA. Residual phenol can be removed from the aqueous phase by extraction with an equal volume of chloroform or ether. After extraction, DNA is usually precipitated with ammonium acetate and ethanol as described in another protocol on this server. Ref. Karger, B. D. (1989) FOCUS 11, 14.

A good source of general information on the properties of phenol can be found in Wallace, Donald M. “Large and Small-Scale Phenol Extractions”. Methods in Enz. Volume 152 guide to Molecular Cloning Techniques. 1987. Academic Press, Inc. Berger and Kimmel, eds. Chap.4, pg 33-41.

(a) At pH 5 to 6 DNA is selectively retained in the organic phase and interphase, leaving RNA in the aqueous phase. Therefore a pH greater than 7 is needed if DNA is to be extracted.

(b) At pH values below 7.6, poly A+ RNA is lost to the organic phase if chloroform is not present.

(c) Optimal RNA yields in phenol extraction are obtained if the salt concentration is less than 0.15 M NaCl. Salt concentration in the sample is not a factor for larger DNA molecules.

To store RNA after extraction use DEPC-treated water.

What is the recommended protocol for phenol-extraction removal of proteins from nucleic acid containing solutions?

Below is a commonly used protocol:

(1) Add an equal volume of buffer-saturated phenol or phenol:chloroform:isoamyl alcohol (25:24:1, v/v/v) to an aqueous nucleic acid solution. Note: for RNA solutions, acid-phenol is recommended.

(2) Vortex, and centrifuge at 14,000 x g for 1 min to separate the phases.

(3) Residual phenol can be removed from the aqueous phase by extraction with an equal volume of chloroform or ether.

(4) After extraction, DNA/RNA is usually precipitated with ammonium acetate and ethanol.

When using a proofreading polymerase such as Pfx, ThermalAce, Pfu, or Vent, is it necessary to neutralize or remove the enzyme after regenerating A-overhangs with a Taq polymerase?

It is best practice to remove the enzymes via phenol-chloroform extraction to prevent the proofreading enzyme from removing the A-overhang again after Taq incubation. If not done, TA cloning efficiencies could be 4-10 fold lower. Alternatively, the PCR product can be cleaned up by gel purification or PCR cleanup column.

What is the optimal pH of the phenol:chloroform mixture for isolation of DNA?

Partitioning of the nucleic acids in phenol is pH dependent. At pH 7.0 or higher, both DNA and RNA partition into the aqueous phase. At an acidic pH (below 7.0) DNA is denatured and will move into the organic phase, but the RNA remains in the aqueous phase. The mixture should be adjusted to at least pH 7.4 for work with DNA.

Recently I came across a DNA purification technique, which uses urea during phenol extraction. What is the purpose of using urea?

Using urea during phenol extraction denatures the protein associated with the DNA and the proteins that bind the genomic DNA to the cell wall.

Citations & References (1)

Citations & References
Abstract
Repair of Sequence-specific 125I-induced Double-strand Breaks by Nonhomologous DNA End Joining in Mammalian Cell-free Extracts.
Authors: Odersky Andrea; Panyutin Irina V; Panyutin Igor G; Schunck Christian; Feldmann Elke; Goedecke Wolfgang; Neumann Ronald D; Obe Gunter; Pfeiffer Petra;
Journal:J Biol Chem
PubMed ID:11821407
In mammalian cells, nonhomologous DNA end joining (NHEJ) is considered the major pathway of double-strand break (DSB) repair. Rejoining of DSB produced by decay of (125)I positioned against a specific target site in plasmid DNA via a triplex-forming oligonucleotide (TFO) was investigated in cell-free extracts from Chinese hamster ovary cells. ... More