2-Ethoxyethanol, 99%, extra pure
2-Ethoxyethanol, 99%, extra pure
2-Ethoxyethanol, 99%, extra pure
Acros Organics

2-Ethoxyethanol, 99%, extra pure

CAS: 110-80-5 | C4H10O2 | 90.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 156020025
also known as 15602
HSN 29094400
Price (INR)
17,847.00
Each
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
17,847.00
Each
Chemical Identifiers
CAS110-80-5
IUPAC Name2-ethoxyethan-1-ol
Molecular FormulaC4H10O2
InChI KeyZNQVEEAIQZEUHB-UHFFFAOYSA-N
SMILESCCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Refractive index1.4060 to 1.4100 (20°C, 589 nm)
Color scale=<10 APHA
GC>=98.5 %
Specific gravity(20°C) 0.928 to 0.932
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only