Ethyl chrysanthemate, 95%, mixture of cis and trans
Ethyl chrysanthemate, 95%, mixture of cis and trans
Ethyl chrysanthemate, 95%, mixture of cis and trans
Thermo Scientific Chemicals

Ethyl chrysanthemate, 95%, mixture of cis and trans

CAS: 97-41-6 | C12H20O2 | 196.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 156061000
Price (EUR)
59,10
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
59,10
Each
Chemical Identifiers
CAS97-41-6
IUPAC Nameethyl 2,2-dimethyl-3-(2-methylprop-1-en-1-yl)cyclopropane-1-carboxylate
Molecular FormulaC12H20O2
InChI KeyVIMXTGUGWLAOFZ-UHFFFAOYNA-N
SMILESCCOC(=O)C1C(C=C(C)C)C1(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.2 %
GC>=94 %
Refractive index1.4590 to 1.4610 (20°C, 589 nm)
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only