Ethyl diethoxyacetate, 98%
Ethyl diethoxyacetate, 98%
Ethyl diethoxyacetate, 98%
Thermo Scientific Chemicals

Ethyl diethoxyacetate, 98%

CAS: 6065-82-3 | C8H16O4 | 176.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 156071000
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS6065-82-3
IUPAC Nameethyl 2,2-diethoxyacetate
Molecular FormulaC8H16O4
InChI KeyXCLBIKIQSCTANZ-UHFFFAOYSA-N
SMILESCCOC(OCC)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Form)Liquid
Refractive index1.4100 to 1.4120 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only