N-Ethylmaleimide, 98+%
N-Ethylmaleimide, 98+%
N-Ethylmaleimide, 98+%
N-Ethylmaleimide, 98+%
Thermo Scientific Chemicals

N-Ethylmaleimide, 98+%

CAS: 128-53-0 | C6H7NO2 | 125.13 g/mol
数量:
5 g
10 g
50 g
250 g
製品番号(カタログ番号) 156100050
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS128-53-0
IUPAC Name1-ethyl-2,5-dihydro-1H-pyrrole-2,5-dione
Molecular FormulaC6H7NO2
InChI KeyHDFGOPSGAURCEO-UHFFFAOYSA-N
SMILESCCN1C(=O)C=CC1=O
さらに表示
Solubility(5% in 100% EtOH) Clear to light turbid,
SolubilityColorless to light yellow solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only