2,2,4,4,6,8,8-Heptamethylnonane, 98%
2,2,4,4,6,8,8-Heptamethylnonane, 98%
2,2,4,4,6,8,8-Heptamethylnonane, 98%
Thermo Scientific Chemicals

2,2,4,4,6,8,8-Heptamethylnonane, 98%

CAS: 4390-04-9 | C16H34 | 226.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1 L
Packaging:
View all
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 156252500
Price (USD)
-
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4390-04-9
IUPAC Name2,2,4,4,6,8,8-heptamethylnonane
Molecular FormulaC16H34
InChI KeyVCLJODPNBNEBKW-UHFFFAOYNA-N
SMILESCC(CC(C)(C)C)CC(C)(C)CC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)Clear colorless
Refractive index1.4380 to 1.4400 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only