HEPES (1 M)
HEPES (1 M)
HEPES (1 M)
Gibco™

HEPES (1 M)

Gibco™ HEPES (N-2-hydroxyethylpiperazine-N-2-ethane sulfonic acid) is a zwitterionic organic chemical buffering agent commonly used in cell culture media. The addition자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
15630080100 mL
1563010620 mL
1563013020 x 100 mL
카탈로그 번호 15630080
제품 가격(KRW)
120,000
온라인 행사
Ends: 31-Dec-2026
133,000
할인액 13,000 (10%)
Each
수량:
100 mL
제품 가격(KRW)
120,000
온라인 행사
Ends: 31-Dec-2026
133,000
할인액 13,000 (10%)
Each
Gibco™ HEPES (N-2-hydroxyethylpiperazine-N-2-ethane sulfonic acid) is a zwitterionic organic chemical buffering agent commonly used in cell culture media. The addition of 10–25 mM HEPES provides extra buffering capacity when cell culture requires extended periods of manipulation outside of a CO2 incubator. HEPES is a good buffering choice for many cell culture systems because it is membrane impermeable, has limited effect on biochemical reactions, is chemically and enzymatically stable, and has very low visible and UV light absorbance.

Dual-site cGMP Manufacturing and Quality System
Gibco™ HEPES is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ HEPES product made in our Grand Island facility (15630-080). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

This product is not used for in vitro diagnostic purpose in some countries.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
사양
CAS7365-45-9
화학물질 이름 또는 재질Zwitterionic Organic Chemical Buffer
색상Clear
제제N-2-Hydroxyethylpiperazine-N-2-Ethane Sulfonic Acid
권장 스토리지Storage conditions: 2°C to 8°C, Shipping conditions: Room temperature, Shelf life: 24 months from date of manufacture
멸균Sterile
멸균 방법Sterile-filtered
농도1 M
용도(애플리케이션)Chromatin Biology
등급Biochemical
분자량(g/mol)238.3 g/mol
물리적 형태Liquid
수량100 mL
용액 유형Buffer
pH7 to 7.5
Unit SizeEach

자주 묻는 질문(FAQ)

Based on the Sodium bicarbonate levels in the medium, what CO2 % is recommended?

If the media formulation contains:

NaHCO3 (g/L) < 1.5, it needs CO2 at 4%;
NaHCO3 (g/L) 1.5 – 2.2, it needs CO2 at 5%;
NaHCO3 (g/L) 2.2 - 3.4, it needs CO2 at 7%;
NaHCO3 (g/L) > 3.5, it needs CO2 at 10% .

*There are some exceptions. Gibco DMEM has always been made according to Dulbecco’s original published formulation, with 3.7 g/L sodium bicarbonate. Customers have been using this medium in CO2 incubators ranging from 5-10% CO2 for decades, usually with no trouble maintaining physiological pH. This also depends on the cell type. Once cells are growing, the pH will drop (due to metabolic accumulation of lactic acid).

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Do high or low levels of sodium bicarbonate affect the pH of my tissue culture media?

Sodium bicarbonate is necessary to control the pH of the solution. To maintain physiological pH, the concentration of the sodium bicarbonate in the medium must be matched with the right level of CO2 in the atmosphere above the medium in the incubator*.

If the sodium bicarbonate is high and the CO2 concentration is low, the pH will become alkaline. This is something that you will see in a bottle of media when it is exposed to air for long periods or when there is a lot of head space in the bottle. The media color will be pinkish or purplish. When this media is put back into a CO2 incubator, the color/pH will change back to the normal orangeish/reddish color or physiological pH.

If the sodium bicarbonate is low and the CO2 is high, the pH will become acidic. This is something you will see when you put medium into a 5% or 10% CO2 incubator.

*There are some exceptions. For instance, Gibco DMEM has always been made according to Dulbecco’s original published formulation, with 3.7g/L sodium bicarbonate. Customers have been using this medium in CO2 incubators ranging from 5-10% CO2 for decades, usually with no trouble maintaining physiological pH. this also depends on the cell type, Once cells are growing, the pH will drop (due to metabolic accumulation of lactic acid).

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

My Gibco HEPES (1M) (Cat. No. 15630080) was accidentally frozen. Can I still use it?

This product should be stored at 2-8 degrees C and should not be frozen. The biggest issue with accidentally freezing this product is its solubility. If this product was accidentally frozen, we recommend placing it in a 2-8 degree environment and allowing it to slowly thaw overnight. If the product is fully thawed and shows no signs of precipitation, then it should still be usable, but we cannot guarantee effectiveness.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Is HEPES (1 M) in distilled water or in saline solution?

1M HEPES is in water with a pH range of 7.2 - 7.5.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the endotoxin specification for HEPES (1 M) (Cat. Nos. 15630080, 15630106, and 15630130)?

Sorry, we do not test endotoxin levels for this product.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

인용 및 참조 문헌 (6)

인용 및 참조 문헌
Abstract
A comparison between different human hepatocyte models reveals profound differences in net glucose production, lipid composition and metabolism in vitro.
Authors:Bonanini F,Singh M,Yang H,Kurek D,Harms AC,Mardinoglu A,Hankemeier T
Journal:Experimental cell research
PubMed ID:38499143
Preparation and characterization of toxic Abeta aggregates for structural and functional studies in Alzheimer's disease research.
Authors:Jan A, Hartley DM, Lashuel HA,
Journal:Nat Protoc
PubMed ID:20539293
'The amyloid cascade hypothesis, supported by strong evidence from genetics, pathology and studies using animal models, implicates amyloid-beta (Abeta) oligomerization and fibrillogenesis as central causative events in the pathogenesis of Alzheimer''s disease (AD). Today, significant efforts in academia, biotechnology and the pharmaceutical industry are devoted to identifying the mechanisms by ... More
Investigating conversion of mechanical force into biochemical signaling in three-dimensional chondrocyte cultures.
Authors:Bougault C, Paumier A, Aubert-Foucher E, Mallein-Gerin F,
Journal:Nat Protoc
PubMed ID:19478808
The culture of chondrocytes embedded within agarose hydrogels maintains chondrocytic phenotype over extended periods and allows analysis of the chondrocyte response to mechanical forces. The mechanisms involved in the transduction of a mechanical stimulus to a physiological process are not completely deciphered. We present protocols to prepare and characterize constructs ... More
Molecular rearrangements of the extracellular vestibule in NMDAR channels during gating.
Authors: Sobolevsky Alexander I; Beck Christine; Wollmuth Lonnie P;
Journal:Neuron
PubMed ID:11779481
Many N-methyl-D-aspartate receptor (NMDAR) channel blockers that have therapeutic potential can be trapped in the closed state. Using a combination of the substituted cysteine accessibility method and open channel blockers, we found that the M3 segment forms the core of the extracellular vestibule, including a deep site for trapping blockers. ... More
Polycystin-1, the product of the polycystic kidney disease 1 gene, co-localizes with desmosomes in MDCK cells.
Authors:Scheffers MS, van der Bent P, Prins F, Spruit L, Breuning MH, Litvinov SV, de Heer E, Peters DJ,
Journal:Hum Mol Genet
PubMed ID:11063733
Polycystin-1 is a novel protein predicted to be a large membrane-spanning glycoprotein with an extracellular N-terminus and an intracellular C-terminus, harboring several structural motifs. To study the subcellular localization, antibodies raised against various domains of polycystin-1 and against specific adhesion complex proteins were used for two-color immunofluorescence staining. In Madine ... More