Product Image
Gibco™

HEPES, 1M Buffer Solution

Gibco™ HEPES (N-2-Hydroxyethylpiperazine-N-2-Ethane Sulfonic Acid) is a zwitterionic organic chemical buffering agent commonly used in cell culture media. The additionRead more
Have Questions?
Catalog NumberQuantity
15630122
also known as 15630-122
20 x 100 mL
Catalog number 15630122
also known as 15630-122
Price (MXN)
-
Quantity:
20 x 100 mL

Gibco™ HEPES (N-2-Hydroxyethylpiperazine-N-2-Ethane Sulfonic Acid) is a zwitterionic organic chemical buffering agent commonly used in cell culture media. The addition of 10 – 25 mM HEPES provides extra buffering capacity when cell culture requires extended periods of manipulation outside of a CO2 incubator. HEPES is a good buffering choice for many cell culture systems because it is membrane impermeable, has limited effect on biochemical reactions, is chemically and enzymatically stable, and has very low visible and UV light absorbance.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Chemical Name or MaterialHEPES Buffer
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Recommended StorageStorage conditions: 2 – 8°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
Shelf Life24 Months
SterilitySterile
Concentration1 M
Physical FormLiquid
Quantity20 x 100 mL
pH7.2 to 7.5
Unit SizeEach