Nunc™ EasYFlask™ Flasks with Nunclon™ Supra Surface
Thermo Scientific™

Nunc™ EasYFlask™ Flasks with Nunclon™ Supra Surface

Experience optimal performance with Thermo Scientific Nunc EasYFlask flasks with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호배양 면적마개 유형
15637225 cm²Filter
15637425 cm²Solid
15637675 cm²Filter
15637875 cm²Solid
156380175 cm²Filter
156382175 cm²Solid
카탈로그 번호 156372
제품 가격(KRW)
395,000
온라인 행사
Ends: 30-Sep-2026
438,000
할인액 43,000 (10%)
Case of 200
배양 면적:
25 cm²
마개 유형:
Filter
제품 가격(KRW)
395,000
온라인 행사
Ends: 30-Sep-2026
438,000
할인액 43,000 (10%)
Case of 200
Experience optimal performance with Thermo Scientific Nunc EasYFlask flasks with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells. The ergonomic design with angled neck and wide opening allows easy access to the growth surface by pipettes or cell scrapers. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.

Thermo Scientific Nunc vessels with Nunclon Supra surface treatment are designed for culturing fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.

The Nunclon Supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Nunclon Supra cultureware is a great xeno-free solution for cell therapy research applications.

Features of Nunc EasYFlask flasks with Nunclon Supra surface include:
• Consistent lot-to-lot performance
• Sterile with SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• Biocompatible per USP Class VI for raw materials
• Shelf life: 5 years

For Laboratory Use
사양
인증/적합성Non-cytotoxic, Non-pyrogenic, USP Class VI
클래스Class VI
마개 유형Filter
설명Nunc EasYFlask, 25 cm²
용도(애플리케이션)General cell culture, cell therapy research, culture of fastidious adherent cells
넥 스타일Angled
케이스당 수200 Flasks
팩당 수10 Flasks
제품 유형Cell Culture Flask
수량200/Cs.
하단 형상Flat
배양 면적25 cm²
물질Polystyrene
멸균Sterile
표면 처리Nunclon Supra
Unit SizeCase of 200