Tribenzylamine, 99+%
Tribenzylamine, 99+%
Tribenzylamine, 99+%
Tribenzylamine, 99+%
Thermo Scientific Chemicals

Tribenzylamine, 99+%

CAS: 620-40-6 | C21H21N | 287.41 g/mol
数量:
100 g
250 g
1 kg
製品番号(カタログ番号) 156472500
価格(JPY)
-
見積もりを依頼する
数量:
250 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS620-40-6
IUPAC Nametribenzylamine
Molecular FormulaC21H21N
InChI KeyMXHTZQSKTCCMFG-UHFFFAOYSA-N
SMILESC(N(CC1=CC=CC=C1)CC1=CC=CC=C1)C1=CC=CC=C1
さらに表示
Appearance (Color)White to cream
GC>=99.0 %
Infrared spectrumConforms
Titration with HClO4>=99.0 % (On anhydrous substance)
Appearance (Form)Powder
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only