Phenethylamine, 99%
Phenethylamine, 99%
Phenethylamine, 99%
Thermo Scientific Chemicals

Phenethylamine, 99%

CAS: 64-04-0 | C8H11N | 121.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
View all
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 156491000
Price (EUR)
22,90
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
22,90
Each
Chemical Identifiers
CAS64-04-0
IUPAC Name2-phenylethan-1-amine
Molecular FormulaC8H11N
InChI KeyBHHGXPLMPWCGHP-UHFFFAOYSA-N
SMILESNCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Refractive index1.5320 to 1.5340 (20°C, 589 nm)
Titration with HClO498.5 to 101.5 %
Appearance (Form)Liquid
Water=<0.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only