Nunc™ EasYFlask™ Cell Culture Flasks
Thermo Scientific™

Nunc™ EasYFlask™ Cell Culture Flasks

Enhance cell attachment, growth, and differentiation with Nunc EasYFlask cell culture treated flasks.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCulture AreaClosure Type
15649975 cm²Filter
15636725 cm²Filter
159910175 cm²Filter
15634025 cm²Solid
159934225 cm²Filter
15647275 cm²Solid
159933225 cm²Solid
159920175 cm²Solid
Catalog number 156499
Price (USD)
496.65
Online exclusive
564.00
Save 67.35
Case of 100
Culture Area:
75 cm²
Closure Type:
Filter
Price (USD)
496.65
Online exclusive
564.00
Save 67.35
Case of 100

Enhance cell attachment, growth, and differentiation with Thermo Scientific Nunc EasYFlask cell culture-treated flasks. These polystyrene flasks receive the proprietary Nunclon Delta surface treatment to maximize adhesion for a broad range of cell types. The ergonomic design with angled neck and wide opening allows easy access to the growth surface by pipettes or cell scrapers.

Features

  • Ergonomic design with a 1/3 turn to open and close the cap
  • Low profile to maximize incubator space
  • Volume graduations printed and molded on both sides
  • Slightly angled side walls for complete viewing under a microscope
  • Sterilized by gamma irradiation and certified to be nonpyrogenic
  • Nunclon Delta Certification, including testing on four different cell types, is provided in every case.

Filter caps

  • Filter membrane provides effective barrier against micro-organisms
  • Well-defined, hydrophobic filter membrane offers continuous venting with consistent gas exchange and no lot-to-lot variation
  • Chemically robust membrane meets USP class VI standards and is produced without animal-derived substances

Solid caps

  • Solid caps feature a Y mark that allows visual verification of vent position
  • Any of the legs in the Y pointing upwards indicates that the cap is in venting position
  • Vent position can be easily distinguished when flasks are stacked in the incubator

Warranty: 90 days

WARNING: Cancer – www.P65Warnings.ca.gov
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceISO 9001:2008, ISO 13485:2003
Closure TypeFilter
Neck StyleAngled
No. per Case100
No. per Pack5
Product TypeFlask
Volume (Metric) Working25 mL
AutoclavableNot Autoclavable
Bottom ShapeFlat
Capacity (Metric)75 cm²
Culture Area75 cm²
DisposableSingle-use
GraduatedYes
MaterialPolystyrene, HDPE
Mouth StyleNarrow
SterilitySterile
Surface TreatmentNunclon Delta
Unit SizeCase of 100

Frequently asked questions (FAQs)

What is the difference between the Nunclon Delta surface and a tissue culture (TC) surface from a competitor?

Our Nunclon Delta surface for adherent cells is a standard tissue culture (TC) surface modification that makes the polystyrene surface more hydrophilic. This facilitates maximum adhesion for a broad range of cell types.

The Nunclon Delta surface is Thermo Scientific Nunclon Delta-certified, which is a Thermo Fisher proprietary certification that has a quality standard to help ensure consistency. Every lot of Thermo Scientific Nunclon Delta-certified cell cultureware is tested with Gibco media for:
- Less than 10% growth variation in L929, Hel299 and primary chick embryo cells
- Less than 15% variation colony formation of V79-4 cells

More information and all applicable products can be found in the "Product selection guide for every step of your cell culture workflow" brochure .

What is the recommended working volume of Nunc adherent cell culture products?

This depends largely on the cell line and working conditions (considerations include headspace for gas exchange, supplements used, and optimal feeding schedule, among others) but we do offer guidelines for development purposes (0.3 to 0.5 mL per cm2) . Note that these volumes are approximate and may have to be modified for best performance. T25 Flask = 7 mL, T75 Flask = 25 mL, T175 Flask = 55 mL, T225 Flask = 70 mL, 300cm2 Flask = 100 mL, 500 cm2 TripleFlask = 200 mL, 35 mm round culture dish = 3 mL, 60 mm round culture dish = 5 mL, 100 mm round culture dish = 12 mL, 150 mm round culture dish = 35 mL, 4 (rectangular) well Multidish = 5 mL, 8 (rectangular) well Multidish = 3 mL, 4 (round) well Multidish = 1 mL, 6 well Multidish = 3 mL, 12 well Multidish = 2 mL, 24 well Multidish = 1 mL, 48 well Multidish = 0.5 mL.

What is the purpose of the Y marking on your Nunc EasYFlask solid caps?

The solid, “vent/close” caps on our EasYFlask culture flasks make use of a unique thread design to allow simple selection of a complete seal or free air exchange with the incubator environment. Once the cap is attached to the flask, any of the legs of the “Y” pointed straight down will indicate that the flask is tightly closed. If the cap is then rotated so that any of the legs point straight up, a channel will open to allow for gas exchange.

What is Nunclon Delta?

Nunclon Delta refers to our performance-certified surface for culture of adherent cells, available in a variety of common formats (flasks, plates, multiwell dishes). It is tested with at least four cell lines (varies by product and manufacturing location) commonly used in research, and it should work well for most robust cell lines. It does not involve addition of coating reagent, but rather is manufactured by exposure of the molded products to a proprietary energy source, which renders the surface more hydrophilic by means of increasing the concentration of oxygen-containing (e.g. hydroxyl) molecular groups. It is by this hydrophilic property that anchorage-dependent cells are able to attach to the surface and become confluent.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (1)

Citations & References
Abstract
A comparison between different human hepatocyte models reveals profound differences in net glucose production, lipid composition and metabolism in vitro.
Authors:Bonanini F,Singh M,Yang H,Kurek D,Harms AC,Mardinoglu A,Hankemeier T
Journal:Experimental cell research
PubMed ID:38499143