3-Methyl-3-buten-1-ol, 97%
3-Methyl-3-buten-1-ol, 97%
3-Methyl-3-buten-1-ol, 97%
Thermo Scientific Chemicals

3-Methyl-3-buten-1-ol, 97%

CAS: 763-32-6 | C5H10O | 86.13 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
產品號碼 156631000
價格 (TWD)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS763-32-6
IUPAC Name3-methylbut-3-en-1-ol
Molecular FormulaC5H10O
InChI KeyCPJRRXSHAYUTGL-UHFFFAOYSA-N
SMILESCC(=C)CCO
檢視更多
規格Specification Sheet規格表
GC>=96.0 %
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4327 to 1.4347 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only