Indoxyl acetate, 97%
Indoxyl acetate, 97%
Indoxyl acetate, 97%
Thermo Scientific Chemicals

Indoxyl acetate, 97%

CAS: 608-08-2 | C10H9NO2 | 175.19 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 156660050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS608-08-2
IUPAC Name1H-indol-3-yl acetate
Molecular FormulaC10H9NO2
InChI KeyJBOPQACSHPPKEP-UHFFFAOYSA-N
SMILESCC(=O)OC1=CNC2=CC=CC=C12
さらに表示
Appearance (Color)White to pink
Infrared spectrumConforms
Melting point127°C to 131°C
GC>=96.0 %
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only