4-(2-Keto-1-benzimidazolinyl)piperidine, 97%
4-(2-Keto-1-benzimidazolinyl)piperidine, 97%
4-(2-Keto-1-benzimidazolinyl)piperidine, 97%
Thermo Scientific Chemicals

4-(2-Keto-1-benzimidazolinyl)piperidine, 97%

CAS: 20662-53-7 | C12H15N3O | 217.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 156700050
Price (USD)
217.65
Special offer
Online exclusive
Ends: 28-Dec-2026
256.00
Save 38.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
217.65
Special offer
Online exclusive
Ends: 28-Dec-2026
256.00
Save 38.35
Each
Chemical Identifiers
CAS20662-53-7
IUPAC Name1-(piperidin-4-yl)-2,3-dihydro-1H-1,3-benzodiazol-2-one
Molecular FormulaC12H15N3O
InChI KeyBYNBAMHAURJNTR-UHFFFAOYSA-N
SMILESO=C1NC2=CC=CC=C2N1C1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to light yellow
Infrared spectrumConforms
Melting point183°C to 185°C
Titration with HClO4>=96.0 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only