Maleimide, 98%
Maleimide, 98%
Maleimide, 98%
Thermo Scientific Chemicals

Maleimide, 98%

CAS: 541-59-3 | C8H4AgKN2O4 | 339.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
25 g
100 g
500 g
Packaging:
View all
Glass bottle
Catalog number 156710250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS541-59-3
IUPAC Namepotassium silver(1+) bis(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-ide)
Molecular FormulaC8H4AgKN2O4
InChI KeyKWLWLQRUFFGWNG-UHFFFAOYSA-L
SMILES[K+].[Ag+].O=C1[N-]C(=O)C=C1.O=C1[N-]C(=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Infrared spectrumConforms
Melting point90°C to 94°C
Titration with NaOH>=97.5 %
Appearance (Color)White to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only