Methyl 3-aminocrotonate, 97%
Methyl 3-aminocrotonate, 97%
Methyl 3-aminocrotonate, 97%
Thermo Scientific Chemicals

Methyl 3-aminocrotonate, 97%

CAS: 14205-39-1 | C5H9NO2 | 115.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 156795000
Price (USD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS14205-39-1
IUPAC Namemethyl (2E)-3-aminobut-2-enoate
Molecular FormulaC5H9NO2
InChI KeyXKORCTIIRYKLLG-ONEGZZNKSA-N
SMILESCOC(=O)\C=C(/C)N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point81°C to 85°C
Appearance (Color)White to yellow
Titration with HClO4>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only