4-Methylaminophenol sulfate, 99%, extra pure
4-Methylaminophenol sulfate, 99%, extra pure
4-Methylaminophenol sulfate, 99%, extra pure
Acros Organics

4-Methylaminophenol sulfate, 99%, extra pure

4-Methylaminophenol sulfate, 99%, C14H18N2O2.H2SO4, CAS Number-55-55-0, Metol, 500g, 13, 441, 100.0, 98.5, Beige to White or Gray, Decomposition Information: >260 deg.C, 1.5770g/mL, 200-237-1, 344.38, Extra Pure, GHS Signal Word: Warning | CAS: 55-55-0 | C14H18N2O2·H2SO4 | 344.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
2.5 kg
Catalog number 156800050
HSN 29222990
Price (INR)
5,356.00
Each
Quantity:
5 g
Request bulk or custom format
Price (INR)
5,356.00
Each
Chemical Identifiers
CAS55-55-0
IUPAC Namebis(4-(methylamino)phenol); sulfuric acid
Molecular FormulaC14H20N2O6S
InChI KeyZVNPWFOVUDMGRP-UHFFFAOYSA-N
SMILESOS(O)(=O)=O.CNC1=CC=C(O)C=C1.CNC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to grey
Heavy metals (as Pb)=<30 ppm
Residue after ignition=<0.15 %
Iron (Fe)=<50 ppm
Appearance (Form)Crystalline powder or crystals
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only