Myristic acid, 99%
Myristic acid, 99%
Myristic acid, 99%
Acros Organics

Myristic acid, 99%

CAS: 544-63-8 | C14H28O2 | 228.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
Packaging:
Glass bottle
Catalog number 156961000
HSN 29159090
Price (INR)
10,220.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
10,220.00
Each
Chemical Identifiers
CAS544-63-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or flakes or chunks or crystalline mass
Infrared spectrumConforms
Appearance (Color)White
GC>=98.5 %
Titration with NaOH>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only