Uracil, ≥ 99 %
Uracil, ≥ 99 %
Uracil, ≥ 99 %
Thermo Scientific Chemicals

Uracil, ≥ 99 %

CAS: 66-22-8 | C4H4N2O2 | 112.09 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
25 g
100 g
500 g
2.5 kg
Verpackung:
Kunststoffflasche
Katalognummer 157305000
Preis (EUR)
373,00
Each
Menge:
500 g
Verpackung:
Kunststoffflasche
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
373,00
Each
Chemikalien-Kennzeichnungen
CAS66-22-8
IUPAC Name1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC4H4N2O2
InChI KeyISAKRJDGNUQOIC-UHFFFAOYSA-N
SMILESO=C1NC=CC(=O)N1
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=99.0 %
Sulfated ash=<0.3 %
Appearance (Color)White to light yellow
Mehr anzeigen
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only