Calconcarboxylic acid, indicator grade
Calconcarboxylic acid, indicator grade
Calconcarboxylic acid, indicator grade
Thermo Scientific Chemicals

Calconcarboxylic acid, indicator grade

Calconcarboxylic acid, indicator grade, Quantity: 250g, Packaging: Glass bottle, Black to Purple, Grade: Extra Pure, Melting Point: 300 deg.C, Molecular Weight: 438.4, CAS: 3737-95-9, EINECS Number: 223-117-, Formula Weight: 438.4, Health Hazard 1: GHS Signal Word: Warning | CAS: 3737-95-9 | C21H14N2O7S | 438.4 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 157312500
Price (USD)
640.65
Special offer
Online exclusive
Ends: 28-Dec-2026
753.00
Save 112.35 (15%)
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
640.65
Special offer
Online exclusive
Ends: 28-Dec-2026
753.00
Save 112.35 (15%)
Each
Chemical Identifiers
CAS3737-95-9
IUPAC Name3-hydroxy-4-{2-[(1Z)-2-oxo-4-sulfo-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}naphthalene-2-carboxylic acid
Molecular FormulaC21H14N2O7S
InChI KeyULIVOAKVRBXKKS-PYCFMQQDSA-N
SMILESOC(=O)C1=C(O)C(N\N=C2/C(=O)C=C(C3=CC=CC=C23)S(O)(=O)=O)=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Suitability(for chel. det. of Ca) Passes test
Appearance (Color)Dark purple to black
Appearance (Form)Powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only