2-Bromophenol, 98%
2-Bromophenol, 98%
2-Bromophenol, 98%
Thermo Scientific Chemicals

2-Bromophenol, 98%

CAS: 95-56-7 | C6H5BrO | 173.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Catalog number 157360500
Price (USD)
-
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS95-56-7
IUPAC Name2-bromophenol
Molecular FormulaC6H5BrO
InChI KeyVADKRMSMGWJZCF-UHFFFAOYSA-N
SMILESOC1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Appearance (Color)Clear colorless to light yellow to light orange
Infrared spectrumConforms
Refractive index1.5870 to 1.5910 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only