Fumaronitrile, 95%
Fumaronitrile, 95%
Fumaronitrile, 95%
Fumaronitrile, 95%
Thermo Scientific Chemicals

Fumaronitrile, 95%

CAS: 764-42-1 | C4H2N2 | 78.06 g/mol
数量:
5 g
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 157401000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS764-42-1
IUPAC Name(2E)-but-2-enedinitrile
Molecular FormulaC4H2N2
InChI KeyKYPOHTVBFVELTG-OWOJBTEDSA-N
SMILESN#C\C=C\C#N
さらに表示
Appearance (Color)White to yellow to light brown
Melting point93°C to 100°C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only