Product Image
Gibco™
Gentamicin (50 mg/mL)
Gentamicin (gentamicin sulfate) is a water-soluble antibiotic drug originally purified from the fungus Micromonospora purpurea. Gentamicin acts by binding toRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
15750094
also known as 15750-094
20 x 20 mL
15750102
also known as 15750-102
20 x 100 mL
Catalog number 15750094
also known as 15750-094
Price (MXN)
-
Quantity:
20 x 20 mL
Gentamicin (gentamicin sulfate) is a water-soluble antibiotic drug originally purified from the fungus Micromonospora purpurea. Gentamicin acts by binding to the 30S subunit of the bacterial ribosome, leading to inhibition of protein synthesis and promotion of cell death in susceptible bacteria.
Using Gentamicin
Gentamicin is effective against a wide variety of gram-positive and gram-negative bacteria, and is used for the prevention of bacterial contamination in cell cultures. The recommended working concentration ranges from 0.5 to 50 μg/mL. We offer a variety of antibiotics and antimycotics for cell culture applications.
Dual-site cGMP manufacturing
Gentamicin is manufactured at a cGMP-compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gentamicin product made in our Grand Island facility (15750-060). This facility is also registered with the FDA as a medical device manufacturer and certified to the ISO 13485 standards.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration50 mg/mL
Sterile FilteredYes
Validated ApplicationPrevention of Cell Culture Contamination
Physical FormLiquid
Quantity20 x 20 mL
TypeGentamicin
Unit SizeEach