1,10-Phenanthroline, 99+%
1,10-Phenanthroline, 99+%
1,10-Phenanthroline, 99+%
Thermo Scientific Chemicals

1,10-Phenanthroline, 99+%

CAS: 66-71-7 | C12H8N2 | 180.21 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 157530250
價格 (TWD)
-
Quantity:
25 g
Request bulk or custom format
化學識別
CAS66-71-7
IUPAC Name1,10-phenanthroline
Molecular FormulaC12H8N2
InChI KeyDGEZNRSVGBDHLK-UHFFFAOYSA-N
SMILESC1=CN=C2C(C=CC3=CC=CN=C23)=C1
檢視更多
規格Specification Sheet規格表
Appearance (Form)Powder
Appearance (Color)White to light yellow to light pink
Infrared spectrumConforms
Melting point114°C to 120°C
Titration with HClO4>=99.0 %
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only