Pyrazinamide, 99%
Pyrazinamide, 99%
Pyrazinamide, 99%
Thermo Scientific Chemicals

Pyrazinamide, 99%

CAS: 98-96-4 | C5H5N3O | 123.11 g/mol
Have Questions?
Catalog NumberQuantityPackaging
15764025025 gPlastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 157640250
Price (USD)
-
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS98-96-4
IUPAC Namepyrazine-2-carboxamide
Molecular FormulaC5H5N3O
InChI KeyIPEHBUMCGVEMRF-UHFFFAOYSA-N
SMILESNC(=O)C1=CN=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White to off-white
Infrared spectrumConforms
Melting point188°C to 191°C
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only