Thermo Scientific™
Ultra-low Temperature Refrigerated/Heated Bath Circulators
This circulator deliver high heating and cooling capacities for rapid heat-up and cool-down times. Fitted with locking castors, drain port, and handles – a perfect fit for any environment.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
With heated tank top designed to avoid ice build up.
  • Effective cooling capacity at ultra low temperatures designed to allow you to reach your specific application temperature requirement.
  • Sealed work area cover.
  • Insulated supply and return ports designed to eliminate ice build up and process temperature variation
Documents
Manuals & protocols