Tetraethyl orthosilicate, 98%
Tetraethyl orthosilicate, 98%
Tetraethyl orthosilicate, 98%
Thermo Scientific Chemicals

Tetraethyl orthosilicate, 98%

CAS: 78-10-4 | C8H20O4Si | 208.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 157810025
Price (USD)
207.65
Online Exclusive
231.00
Save 23.35 (10%)
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
207.65
Online Exclusive
231.00
Save 23.35 (10%)
Each
Chemical Identifiers
CAS78-10-4
IUPAC Nametetraethyl silicate
Molecular FormulaC8H20O4Si
InChI KeyBOTDANWDWHJENH-UHFFFAOYSA-N
SMILESCCO[Si](OCC)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<20 APHA
Refractive index1.3820 to 1.3840 (20°C, 589 nm)
Appearance (Form)Clear liquid
GC>=97.5 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only