4-Methylbenzenesulfonhydrazide, 97%
4-Methylbenzenesulfonhydrazide, 97%
4-Methylbenzenesulfonhydrazide, 97%
Thermo Scientific Chemicals

4-Methylbenzenesulfonhydrazide, 97%

CAS: 1576-35-8 | C7H10N2O2S | 186.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number 157860250
Price (EUR)
28,65
Online exclusive
32,00
Save 3,35 (10%)
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
28,65
Online exclusive
32,00
Save 3,35 (10%)
Each
Chemical Identifiers
CAS1576-35-8
IUPAC Name4-methylbenzene-1-sulfonohydrazide
Molecular FormulaC7H10N2O2S
InChI KeyICGLPKIVTVWCFT-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)S(=O)(=O)NN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
HPLC>=96.0 %
Appearance (Color)White to off-white to light grey
Infrared spectrumConforms
Melting point105°C to 113°C (Dec)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only