2,4,5-Trimethoxybenzaldehyde, 98%
2,4,5-Trimethoxybenzaldehyde, 98%
2,4,5-Trimethoxybenzaldehyde, 98%
Thermo Scientific Chemicals

2,4,5-Trimethoxybenzaldehyde, 98%

CAS: 4460-86-0 | C10H12O4 | 196.20 g/mol
数量:
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 157940250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4460-86-0
IUPAC Name2,4,5-trimethoxybenzaldehyde
Molecular FormulaC10H12O4
InChI KeyIAJBQAYHSQIQRE-UHFFFAOYSA-N
SMILESCOC1=CC(OC)=C(C=O)C=C1OC
さらに表示
Appearance (Color)White to light beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point111°C to 115°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only