Anisonitrile, 99%
Anisonitrile, 99%
Anisonitrile, 99%
Thermo Scientific Chemicals

Anisonitrile, 99%

CAS: 874-90-8 | C8H7NO | 133.15 g/mol
数量:
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 158050250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS874-90-8
IUPAC Name4-methoxybenzonitrile
Molecular FormulaC8H7NO
InChI KeyXDJAAZYHCCRJOK-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)C#N
さらに表示
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point57°C to 60°C
GC>=98.5 %
Appearance (Color)White to light beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only