3,5-Dimethoxyphenol, 97%
3,5-Dimethoxyphenol, 97%
3,5-Dimethoxyphenol, 97%
Thermo Scientific Chemicals

3,5-Dimethoxyphenol, 97%

CAS: 500-99-2 | C8H10O3 | 154.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 158120100
Price (USD)
86.65
Special offer
Online exclusive
Ends: 28-Dec-2026
102.00
Save 15.35
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
86.65
Special offer
Online exclusive
Ends: 28-Dec-2026
102.00
Save 15.35
Each
Chemical Identifiers
CAS500-99-2
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Melting point40°C to 46°C
Appearance (Color)White to yellow to pink
Appearance (Form)Crystalline powder or crystals or crystalline mass
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only