Chloroform, 99+%, extra pure, stabilized with ethanol
Chloroform, 99+%, extra pure, stabilized with ethanol
Chloroform, 99+%, extra pure, stabilized with ethanol
Thermo Scientific Chemicals

Chloroform, 99+%, extra pure, stabilized with ethanol

CAS: 67-66-3 | CHCl3 | 119.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
5 L
Packaging:
View all
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 158210025
Price (TWD)
-
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS67-66-3
IUPAC Nametrichloromethane
Molecular FormulaCHCl3
InChI KeyHEDRZPFGACZZDS-UHFFFAOYSA-N
SMILESClC(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Infrared spectrumConforms
Water=<0.02 % (K.F.)
Stabilizer0.5 to 1.0 % ethanol
Color scale=<25 APHA
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only