4,4'-Isopropylidenediphenol, 97%
4,4'-Isopropylidenediphenol, 97%
4,4'-Isopropylidenediphenol, 97%
Thermo Scientific Chemicals

4,4'-Isopropylidenediphenol, 97%

CAS: 80-05-7 | C15H16O2 | 228.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 158240020
Price (EUR)
86,65
Online exclusive
96,40
Save 9,75 (10%)
Each
Quantity:
2 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
86,65
Online exclusive
96,40
Save 9,75 (10%)
Each
Chemical Identifiers
CAS80-05-7
IUPAC Name4-[2-(4-hydroxyphenyl)propan-2-yl]phenol
Molecular FormulaC15H16O2
InChI KeyIISBACLAFKSPIT-UHFFFAOYSA-N
SMILESCC(C)(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Flakes or prills
GC>=96.0 %
Infrared spectrumConforms
Melting point155°C to 160°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only