Trichloroethylene, 99+%, extra pure, stabilized
Trichloroethylene, 99+%, extra pure, stabilized
Trichloroethylene, 99+%, extra pure, stabilized
Thermo Scientific Chemicals

Trichloroethylene, 99+%, extra pure, stabilized

CAS: 79-01-6 | C2HCl3 | 131.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 158310025
Price (USD)
-
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS79-01-6
IUPAC Name1,1,2-trichloroethene
Molecular FormulaC2HCl3
InChI KeyXSTXAVWGXDQKEL-UHFFFAOYSA-N
SMILESClC=C(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Residue after evaporation=<0.002 %
StabilizerStabilized
Color scale=<15 APHA
Infrared spectrumConforms
Refractive index1.4760 to 1.4780 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only