n-Butilo isocianida, +98 %, Thermo Scientific Chemicals
n-Butilo isocianida, +98 %, Thermo Scientific Chemicals
n-Butilo isocianida, +98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Butilo isocianida, +98 %, Thermo Scientific Chemicals

CAS: 2769-64-4 | C5H9N | 83.13 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
Envase:
Frasco de vidrio
Número de catálogo 158380050
Precio (EUR)
265,00
Each
Cantidad:
5 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Precio (EUR)
265,00
Each
Identificadores químicos
CAS2769-64-4
IUPAC Name1-isocyanobutane
Molecular FormulaC5H9N
InChI KeyFSBLVBBRXSCOKU-UHFFFAOYSA-N
SMILESCCCC[N+]#[C-]
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Form)Liquid
GC>=98.0 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.3950 to 1.3970 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only