Cyclohexyl isocyanide, 99%
Cyclohexyl isocyanide, 99%
Cyclohexyl isocyanide, 99%
Thermo Scientific Chemicals

Cyclohexyl isocyanide, 99%

CAS: 931-53-3 | C7H11N | 109.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 158400050
Price (EUR)
97,65
Online exclusive
108,00
Save 10,35 (10%)
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
97,65
Online exclusive
108,00
Save 10,35 (10%)
Each
Chemical Identifiers
CAS931-53-3
IUPAC Nameisocyanocyclohexane
Molecular FormulaC7H11N
InChI KeyXYZMOVWWVXBHDP-UHFFFAOYSA-N
SMILES[C-]#[N+]C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light brown
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.0 %
Refractive index1.4490 to 1.4520 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only