Aniline hydrochloride, 99%
Aniline hydrochloride, 99%
Aniline hydrochloride, 99%
Thermo Scientific Chemicals

Aniline hydrochloride, 99%

CAS: 142-04-1 | C6H8ClN | 129.59 g/mol
数量:
100 g
500 g
2.5 kg
パッケージ:
Plastic bottle
製品番号(カタログ番号) 158410025
価格(JPY)
-
数量:
2.5 kg
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS142-04-1
IUPAC Namehydrogen aniline chloride
Molecular FormulaC6H8ClN
InChI KeyMMCPOSDMTGQNKG-UHFFFAOYSA-N
SMILES[H+].[Cl-].NC1=CC=CC=C1
さらに表示
Appearance (Color)White to grey
Infrared spectrumConforms
Titration with HClO4>=98.5 %
Appearance (Form)Powder or crystals or flakes
Melting point196°C to 200°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only