Aniline hydrochloride, 99%
Aniline hydrochloride, 99%
Aniline hydrochloride, 99%
Thermo Scientific Chemicals

Aniline hydrochloride, 99%

CAS: 142-04-1 | C6H8ClN | 129.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 158415000
Price (EUR)
64,40
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
64,40
Each
Chemical Identifiers
CAS142-04-1
IUPAC Namehydrogen aniline chloride
Molecular FormulaC6H8ClN
InChI KeyMMCPOSDMTGQNKG-UHFFFAOYSA-N
SMILES[H+].[Cl-].NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to grey
Infrared spectrumConforms
Titration with HClO4>=98.5 %
Appearance (Form)Powder or crystals or flakes
Melting point196°C to 200°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only