4-Amino-1-naphthol hydrochloride, 90%, technical
4-Amino-1-naphthol hydrochloride, 90%, technical
4-Amino-1-naphthol hydrochloride, 90%, technical
4-Amino-1-naphthol hydrochloride, 90%, technical
Thermo Scientific Chemicals

4-Amino-1-naphthol hydrochloride, 90%, technical

CAS: 5959-56-8 | C10H9NO·HCl | 195.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 158510250
Price (EUR)
247,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
247,00
Each
Add to cart
Chemical Identifiers
CAS5959-56-8
IUPAC Name4-hydroxynaphthalen-1-aminium chloride
Molecular FormulaC10H10ClNO
InChI KeyFDBQTRARWCKEJY-UHFFFAOYSA-N
SMILES[Cl-].[NH3+]C1=C2C=CC=CC2=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pink to grey
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=88 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only