4-Chlorobenzenesulfonyl chloride, 97%
4-Chlorobenzenesulfonyl chloride, 97%
4-Chlorobenzenesulfonyl chloride, 97%
Thermo Scientific Chemicals

4-Chlorobenzenesulfonyl chloride, 97%

CAS: 98-60-2 | C6H4Cl2O2S | 211.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 158561000
Price (USD)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS98-60-2
IUPAC Name4-chlorobenzene-1-sulfonyl chloride
Molecular FormulaC6H4Cl2O2S
InChI KeyZLYBFBAHAQEEQQ-UHFFFAOYSA-N
SMILESClC1=CC=C(C=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals and/or chunks
Melting point50°C to 55°C
Appearance (Color)White to light yellow
Titration Argentometric96.0 to 104.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only