trans-Cinnamic Acid, 98+%
trans-Cinnamic Acid, 98+%
trans-Cinnamic Acid, 98+%
Thermo Scientific Chemicals

trans-Cinnamic Acid, 98+%

CAS: 140-10-3 | C9H8O2 | 148.161 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 158575000
Price (USD)
99.65
Special offer
Online exclusive
Ends: 30-Jun-2026
117.00
Save 17.35 (15%)
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
99.65
Special offer
Online exclusive
Ends: 30-Jun-2026
117.00
Save 17.35 (15%)
Each
Chemical Identifiers
CAS140-10-3
IUPAC Name(2E)-3-phenylprop-2-enoic acid
Molecular FormulaC9H8O2
InChI KeyWBYWAXJHAXSJNI-VOTSOKGWSA-N
SMILESOC(=O)\C=C\C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with NaOH>=98.0 %
Melting point131°C to 136°C
Appearance (Form)Crystalline powder
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only