2-Hydroxyhippuric acid, 95%
2-Hydroxyhippuric acid, 95%
2-Hydroxyhippuric acid, 95%
Thermo Scientific Chemicals

2-Hydroxyhippuric acid, 95%

CAS: 487-54-7 | C9H9NO4 | 195.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Catalog number 158620025
Price (USD)
-
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS487-54-7
IUPAC Name2-[(2-hydroxyphenyl)formamido]acetic acid
Molecular FormulaC9H9NO4
InChI KeyONJSZLXSECQROL-UHFFFAOYSA-N
SMILESOC(=O)CNC(=O)C1=CC=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
Melting point163°C to 168°C
Appearance (Color)White to cream
HPLC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only